Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKHD1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals BC11060480AF350
Description
PKHD1 Polyclonal specifically detects PKHD1 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Peptide ELISA.Specifications
PKHD1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence, Peptide ELISA | |
polycystic kidney and hepatic disease 1 (autosomal recessive) | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse | |
IgG |
Immunocytochemistry, Immunofluorescence, ELISA | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
PKHD1 | |
A synthetic peptide derived from the human PKHD1 protein (within residues 3800-3900). [UniProtNo. Q8TCZ9] | |
0.1 mL | |
5314.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction