Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKHD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556682
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556682 Supplier Invitrogen™ Supplier No. PA556682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KTTTVNYVRD TLSNPRGWMA LLLDQETYSL QSENLWINRS LQYSATFDNF APGNYLLLVH TDLPPYPDIL LRCGSRVGLS FPFLPSPGQN Q Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 26%.

The protein encoded by this gene is predicted to have a single transmembrane (TM)-spanning domain and multiple copies of an immunoglobulin-like plexin-transcription-factor domain. Alternative splicing results in two transcript variants encoding different isoforms. Other alternatively spliced transcripts have been described, but the full length sequences have not been determined. Several of these transcripts are predicted to encode truncated products which lack the TM and may be secreted. Mutations in this gene cause autosomal recessive polycystic kidney disease, also known as polycystic kidney and hepatic disease-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKHD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PKHD1
Gene Accession No. P08F94
Gene Alias AI118496; AI182499; ARPKD; FCYT; Fibrocystin; PKHD1; polycystic kidney and hepatic disease 1 (autosomal recessive); polycystic kidney and hepatic disease 1 protein; Polyductin; TIG multiple domains 1; TIGM1; Tigmin
Gene Symbols PKHD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PKHD1. Recombinant protein control fragment (Product #RP-94948).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5314
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.