Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pkhd1 Rat anti-Human, Clone:8G12A1, Abnova™

Catalog No. 89333107 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-107 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-107 Supplier Abnova Corporation Supplier No. MAB16657
Add to Cart
Add to Cart

Rat monoclonal antibody raised against recombinant mouse Pkhd1.

Specifications

Antigen Pkhd1
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8G12A1
Conjugate Unconjugated
Description polycystic kidney and hepatic disease 1
Dilution ELISA (1:10000)
Immunohistochemistry (1:200-1:1000)
Immunocytochemistry (1:200-1:1000)
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene Pkhd1
Gene Alias AI118496, AI182499, FPC, Tigm1
Host Species Rat
Immunogen Recombinant protein corresponding to amino acid 3878-4060 of mouse Pkhd1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 241035
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.