Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKIA Mouse anti-Human, Clone: OTI1G5, TrueMAB™, Origene

Mouse Monoclonal Antibody
Supplier: OriGene TA810311S

This item is not returnable.
View return policy
Description
PKIA Monoclonal antibody specifically detects PKIA in Human samples. It is validated for Western BlotSpecifications
PKIA | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P61925 | |
Mouse | |
Affinity chromatography | |
RUO | |
5569 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI1G5 | |
Unconjugated | |
PKIA | |
AI415001; cAMP-dependent protein kinase inhibitor alpha; cAMP-dependent protein kinase inhibitor, muscle/brain isoform; PKIA; PKIalpha; PKI-alpha; PRKACN1; protein kinase (cAMP-dependent, catalytic) inhibitor alpha; protein kinase inhibitor, alpha | |
Full length human recombinant protein of human PKIA produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction