Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKIB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144029
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144029 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144029 Supplier Invitrogen™ Supplier No. PA5144029
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - IHC: human placenta tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This gene encodes a member of the cAMP-dependent protein kinase inhibitor family. The encoded protein may play a role in the protein kinase A (PKA) pathway by interacting with the catalytic subunit of PKA, and overexpression of this gene may play a role in prostate cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Jul 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKIB
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene PKIB
Gene Accession No. Q9C010
Gene Alias cAMP-dependent protein kinase (catalytic subunit binding) inhibtor 2; cAMP-dependent protein kinase inhibitor 2; cAMP-dependent protein kinase inhibitor beta; cAMP-dependent protein kinase inhibitor bet-like protein; cAMP-dependent protein kinase inhibitor, testis isoform; im:7054531; Pkib; Pkibeta; PKI-beta; PKINH; PRKACN2; protein kinase (cAMP dependent, catalytic) inhibitor beta; protein kinase (cAMP-dependent, catalytic) inhibitor beta; protein kinase inhibitor beta, (cAMP-dependent, catalytic) inhibitor beta; protein kinase inhibitor beta, cAMP dependent, catalytic; protein kinase inhibitor beta, cAMP dependent, testis specific; protein kinase inhibitor, beta; protein kinase, cAMP-dependent, catalytic, inhibitor 2; RATPKINH; si:dkey-150h13.1
Gene Symbols PKIB
Host Species Rabbit
Immunogen E.coli-derived human PKIB recombinant protein (Position: M1-K78).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5570
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.