Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKLR Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297071H
Description
PKLR Polyclonal specifically detects PKLR in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PKLR | |
Polyclonal | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 2.7.1.40, PK1PKRL, PKL, PKR, Pyruvate kinase 1, pyruvate kinase isozyme R/L, pyruvate kinase isozymes R/L, pyruvate kinase type L, pyruvate kinase, liver and blood cell, pyruvate kinase, liver and RBC, Red cell/liver pyruvate kinase, RPK, R-type/L-type pyruvate kinase | |
Produced in rabbits immunized with purified, recombinant PKLR (NP_000289.1; Leu47-Ser574). | |
0.1 mL | |
Chaperone Mediated Autophagy (CMA), Lipid and Metabolism, Signal Transduction | |
5313 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction