Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKLR Monoclonal Antibody (OTI1G9), TrueMAB™, OriGene
SDP

Catalog No. 502542300
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2300 30 μL
50-167-2560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2300 Supplier OriGene Supplier No. TA501418S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PKLR Monoclonal antibody specifically detects PKLR in Human samples. It is validated for Western Blot

Specifications

Antigen PKLR
Applications Western Blot
Classification Monoclonal
Clone OTI1G9
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PKLR
Gene Accession No. P30613
Gene Alias isozymes R/L; L-PK; PK1; Pk-1; PKL; Pklg; Pklr; PKR; PKRL; Pyruvate kinase 1; pyruvate kinase isozyme R/L; pyruvate kinase isozymes L/R; pyruvate kinase isozymes R/L; pyruvate kinase liver and red blood cell; pyruvate kinase PKLR; pyruvate kinase type L; pyruvate kinase, liver and blood cell; pyruvate kinase, liver and RBC; pyruvate kinase, liver and red blood cell; red cell/liver pyruvate kinase; RPK; R-PK; R-type/L-type pyruvate kinase
Host Species Mouse
Immunogen Full length human recombinant protein of human PKLR produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5313
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.