Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKLR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588337
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588337 Supplier Invitrogen™ Supplier No. PA588337
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLSRDPTEVT AIGAVEAAFK CCAAAIIVLT TTGRSAQLLS RYRPRAAVIA VTRSAQAARQ VHLCRGVFPL LYREPPEAIW ADDVDRRVQF GIESGKLRGF LRVGDLVIVV T; Positive Samples: BxPC-3, HepG2, 293T, Mouse kideny, Mouse liver, Mouse heart, Rat kidney.

The protein encoded by this gene is a pyruvate kinase that catalyzes the transphosphorylation of phohsphoenolpyruvate into pyruvate and ATP, which is the rate-limiting step of glycolysis. Defects in this enzyme, due to gene mutations or genetic variations, are the common cause of chronic hereditary nonspherocytic hemolytic anemia (CNSHA or HNSHA). Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKLR
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PKLR
Gene Accession No. P12928, P30613, P53657
Gene Alias isozymes R/L; L-PK; PK1; Pk-1; PKL; Pklg; Pklr; PKR; PKRL; Pyruvate kinase 1; pyruvate kinase isozyme R/L; pyruvate kinase isozymes L/R; pyruvate kinase isozymes R/L; pyruvate kinase liver and red blood cell; pyruvate kinase PKLR; pyruvate kinase type L; pyruvate kinase, liver and blood cell; pyruvate kinase, liver and RBC; pyruvate kinase, liver and red blood cell; red cell/liver pyruvate kinase; RPK; R-PK; R-type/L-type pyruvate kinase
Gene Symbols PKLR
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 446-556 of human PKLR (NP_0002891).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18770, 24651, 5313
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.