Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKM1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP214833AF647
Description
PKM1 Polyclonal specifically detects PKM1 in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
PKM1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting | |
PK1, Pyruvate kinase isozymes M1, Pyruvate kinase M1 | |
Rabbit | |
Affinity Purified | |
RUO | |
This antibody is specific for the M1 isoform of PKM. | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PKM | |
A synthetic peptide made to a C-terminal portion of the human PKM1 protein (between residues 350-450) [UniProt P14618-2]. | |
0.1 mL | |
Primary | |
Human, Mouse, Bovine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction