Learn More
Invitrogen™ PKM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120660
Description
Positive test controls include: Mouse skeletal muscle, Mouse heart, Rat skeletal muscle. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: GSDVANAVLD GADCIMLSGE TAKGDYPLEA VRMQHLIARE AEAAMFHRKL FEELVRASSH STDLMEAMAM GSVEASYKCL AAALIVLTES GRSAHQVARY R.
PKM1 encodes a protein involved in glycolysis. The encoded protein is a pyruvate kinase that catalyzes the transfer of a phosphoryl group from phosphoenolpyruvate to ADP, generating ATP and pyruvate. This protein has been shown to interact with thyroid hormone and may mediate cellular metabolic effects induced by thyroid hormones. This protein has been found to bind Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, suggesting a role of this protein in bacterial pathogenesis. Several alternatively spliced transcript variants encoding a few distinct isoforms have been reported.
Specifications
PKM1 | |
Polyclonal | |
Unconjugated | |
PKM | |
AA414905; AL024370; AL024424; CTHBP; Cytosolic thyroid hormone-binding protein; epididymis secretory protein Li 30; HEL-S-30; OIP3; OIP-3; Opa-interacting protein 3; p58; PK, muscle type; PK2; Pk-2; PK3; Pk-3; Pkm; PKM1; PKM12; PKM2; Pykm; Pyruvate kinase 2/3; pyruvate kinase 3; pyruvate kinase isozymes M1/M2; pyruvate kinase muscle isozyme; Pyruvate kinase PKM; pyruvate kinase, muscle; TCB; THBP1; Thyroid hormone-binding protein 1; thyroid hormone-binding protein, cytosolic; tumor M2-PK | |
Rabbit | |
Affinity Chromatography | |
RUO | |
18746, 25630, 5315 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.82 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P11980, P14618, P52480 | |
PKM | |
A synthetic peptide corresponding to a sequence within amino acids 350-450 of human PKM1-specific (NP_001193728.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.