Learn More
Invitrogen™ PKM2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589522
Description
Immunogen sequence: SEAGTAFIQT QQLHAAMADT FLEHMCRLDI DSPPITARNT GIICTIGPAS RSVETLKEMI KSGMNVARLN FSHGTHEYHA ETIKNVRTAT ESFASDPILY RPVAVALDTK GPEIRTGLIK GSGTAEVELK KGATLKITLD NAYMEKCDEN ILWLDYKNIC KVVEVGSKIY VDDGLISLQV KQKGADFLVT EVENGGSLGS KKGVNLPGAA V Positive Samples: U-251MG, LO2, HeLa, A-549, Mouse kidney, Mouse brain, Mouse lung, Rat brain; Cellular Location: Cytoplasm, Nucleus.
PKM2 is a pyruvate kinase that catalyzes the production of phosphoenolpyruvate from pyruvate and ATP. PKM2 has been shown to interact with thyroid hormone, and thus may mediate cellular metabolic effects induced by thyroid hormones. PKM2 has been found to bind Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, suggesting a role of this protein in bacterial pathogenesis. Three alternatively spliced transcript variants encoding two distinct isoforms of PKM2 have been reported.
Specifications
PKM2 | |
Polyclonal | |
Unconjugated | |
PKM | |
AA414905; AL024370; AL024424; ATP:pyruvate 2-o-phosphotransferase; CTHBP; Cytosolic thyroid hormone-binding protein; epididymis secretory protein Li 30; HEL-S-30; isozymes M1/M2; m2pk; muscle pyruvate kinase; OIP3; OIP-3; OPA-interacting protein 3; p58; PK, muscle type; PK2; Pk-2; Pk3; Pk-3; PKLR; PKM; pkm.L; PKM12; PKM2; Pykm; pyruvate kinase; pyruvate kinase 2/3; pyruvate kinase 3; pyruvate kinase isozymes M1/M2; pyruvate kinase M1/2; pyruvate kinase M1/2 L homeolog; pyruvate kinase M1/2 L homeolog; pyruvate kinase, muscle; pyruvate kinase muscle isozyme; Pyruvate kinase PKM; pyruvate kinase, liver and RBC; pyruvate kinase, muscle; TCB; THBP1; thyroid hormone-binding protein 1; thyroid hormone-binding protein, cytosolic; tu; tumor M2-PK; XELAEV_18018020mg | |
Rabbit | |
Affinity chromatography | |
RUO | |
18746, 25630, 5315 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P11980, P14618, P52480 | |
PKM | |
Recombinant fusion protein containing a sequence corresponding to amino acids 11-221 of PKM (NP_0011937281). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.