Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKMYT1 Antibody (OTI5E1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP202275
Description
PKMYT1 Monoclonal specifically detects PKMYT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| PKMYT1 | |
| Monoclonal | |
| 0.69 mg/mL | |
| Western Blot 1:500, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
| PKMYT1 | |
| Full length human recombinant protein of human PKMYT1 (NP_004194) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Monkey | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI5E1 | |
| Unconjugated | |
| DKFZp547K1610, EC 2.7.11.1, FLJ20093, Myt1 kinase, MYT1membrane-associated tyrosine- and threonine-specific cdc2-inhibitory kinase, protein kinase, membrane associated tyrosine/threonine 1 | |
| Mouse | |
| 54.3 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 9088 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction