Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKMYT1 Monoclonal Antibody (OTI2B4)
GREENER_CHOICE

Catalog No. PIMA525029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525029 Supplier Invitrogen™ Supplier No. MA525029
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PKMYT1 Monoclonal Antibody for Flow, ICC/IF, Western Blot

PKMYT1 serine/threonine kinase is a member of a family of neural specific, zinc finger-containing DNA-binding proteins which binds to the promoter regions of proteolipid proteins of the central nervous system and plays a role in the developing nervous system. MYT1 of the oligodendrocyte lineage, along with a closely related CCHC zinc finger, is expressed in developing neurons in the mammalian central nervous system. The recombinant PKMYT1 fragments containing either the upstream 2 zinc fingers or the downstream 4 zinc fingers bound the same cis regulatory element in the PLP1 promoter in vitro and DNA binding required Zn2+, but not other divalent cations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKMYT1
Applications Flow Cytometry, Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2B4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Pkmyt1
Gene Accession No. Q99640
Gene Alias 6230424P17; AW209059; Membrane-associated tyrosine- and threonine-specific cdc2-inhibitory kinase; membrane-associated tyrosine-and threonine-specific cdc2-inhibitory kinase; Myt1; Myt1 kinase; Pkmyt1; pkmyt1.S; PPP1R126; protein kinase, membrane associated tyrosine/threonine 1; protein kinase, membrane associated tyrosine/threonine 1 S homeolog; protein phosphatase 1, regulatory subunit 126; RGD1305434; XELAEV_18047881mg
Gene Symbols Pkmyt1
Host Species Mouse
Immunogen Full length human recombinant protein of PKMYT1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9088
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.