Learn More
Invitrogen™ PKMYT1 Monoclonal Antibody (OTI5E1)

Description
PKMYT1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | PKMYT1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI5E1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Pkmyt1 |
| Gene Accession No. | Q99640 |
| Gene Alias | 6230424P17; AW209059; Membrane-associated tyrosine- and threonine-specific cdc2-inhibitory kinase; membrane-associated tyrosine-and threonine-specific cdc2-inhibitory kinase; Myt1; Myt1 kinase; Pkmyt1; pkmyt1.S; PPP1R126; protein kinase, membrane associated tyrosine/threonine 1; protein kinase, membrane associated tyrosine/threonine 1 S homeolog; protein phosphatase 1, regulatory subunit 126; RGD1305434; XELAEV_18047881mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.