Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKN1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298761MFV450
Description
PKN1 Polyclonal antibody specifically detects PKN1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
PKN1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
DBK, EC 2.7.11, MGC46204, PAK1Serine-threonine protein kinase N, PKNPKN-ALPHA, PRK1PAK-1, PRKCL1protein kinase C-related kinase 1, Protease-activated kinase 1, Protein kinase C-like 1EC 2.7.11.13, Protein kinase C-like PKN, protein kinase N1, Protein kinase PKN-alpha, Protein-kinase C-related kinase 1, serine/threonine protein kinase N, serine/threonine-protein kinase N1, serine-threonine kinase N | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human PKN1. | |
0.1 mL | |
Protein Kinase | |
5585 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction