Learn More
Invitrogen™ PKN1 Monoclonal Antibody (1B10)

Description
Peptide Sequence: LDMEPQGCLV AEVTFRNPVI ERIPRLRRQK KIFSKQQGKA FQRARQMNID VATWVRLLRR LIPNATGTGT FSPGASPGSE ARTTGDISVE KLNLGTDSDS SPQKSSRDPP SSPSSLSSPI QESTAPELPS ETQETPGPAL CSPLRKSPLT LEDF.
Specifications
Specifications
| Antigen | PKN1 |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1B10 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | PKN1 |
| Gene Accession No. | Q16512 |
| Gene Alias | DBK; F730027O18Rik; PAK1; PAK-1; PKN; PKN1; PKN-ALPHA; PRK1; Prkcl1; Protease-activated kinase 1; protein kinase C-like 1; Protein kinase C-like PKN; protein kinase C-related kinase 1; protein kinase N1; protein kinase PKN; protein kinase PKN-alpha; Protein-kinase C-related kinase 1; serine/threonine kinase 3; serine/threonine protein kinase N; serine/threonine-protein kinase N1; serine-threonine kinase N; serine-threonine protein kinase N; Stk3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.