Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PKN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552036
Description
Immunogen sequence: QKLGLLREAL ERRLGELPAD HPKGRLLREE LAAASSAAFS TRLAGPFPAT HYSTLCKPAP LTGTLEVRVV GCRDLPETIP WNPTPSMGGP GTPDSRPPFL SRPARGLYSR SGSLSGRSSL KAEAENTSEV S Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.
PKN1 (PRK1) belongs to the protein kinase C superfamily. This kinase is activated by the Rho family of small G proteins and may mediate the Rho-dependent signaling pathway. This kinase can also be activated by phospholipids. The 3-phosphoinositide dependent protein kinase-1 (PDPK1/PDK1) is reported to phosphorylate this kinase, which may mediate insulin signals to the actin cytoskeleton. The proteolytic activation of this kinase by caspase-3 or related proteases during apoptosis suggests its role in signal transduction related to apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
PKN1 | |
Polyclonal | |
Unconjugated | |
PKN1 | |
DBK; F730027O18Rik; PAK1; PAK-1; PKN; PKN1; PKN-ALPHA; PRK1; Prkcl1; Protease-activated kinase 1; protein kinase C-like 1; Protein kinase C-like PKN; protein kinase C-related kinase 1; protein kinase N1; protein kinase PKN; protein kinase PKN-alpha; Protein-kinase C-related kinase 1; serine/threonine kinase 3; serine/threonine protein kinase N; serine/threonine-protein kinase N1; serine-threonine kinase N; serine-threonine protein kinase N; Stk3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29355, 320795, 5585 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P70268, Q16512, Q63433 | |
PKN1 | |
Recombinant protein corresponding to Human PKN1. Recombinant protein control fragment (Product #RP-102277). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction