Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKN2 Antibody (1D1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237573
Description
PKN2 Monoclonal specifically detects PKN2 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PKN2 | |
Monoclonal | |
Unconjugated | |
Ascites with 0.03% Sodium Azide | |
PKN2 | |
Purified recombinant fragment of human PRK2 expressed in E. Coli. | |
Unpurified | |
RUO | |
Primary | |
Human, Mouse, Rat, Monkey, Primate | |
Ascites |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1D1 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin | |
cardiolipin-activated protein kinase Pak2, EC 2.7.11, EC 2.7.11.13, Pak-2, PAK2, PRK2MGC150606, PRKCL2PRO2042, Protein kinase C-like 2MGC71074, protein kinase N2, Protein-kinase C-related kinase 2, serine/threonine-protein kinase N2 | |
Mouse | |
140 kDa | |
0.1 mL | |
Protein Kinase | |
5586 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction