Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKN2 Recombinant Rabbit Monoclonal Antibody (11H3)
GREENER_CHOICE

Catalog No. FEMA550169
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550169 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550169 Supplier Invitrogen™ Supplier No. MA550169
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

PKN2 Recombinant Monoclonal Antibody for Western Blot, Flow, ELISA

PKN2 (PRK2), a close relative of the PKCs, is a regulator of mitosis and exit from cytokinesis. Plays a role in the regulation of cell cycle progression, actin cytoskeleton assembly, cell migration, cell adhesion, tumor cell invasion and transcription activation signaling processes. Phosphorylates CTTN in hyaluronan-induced astrocytes and hence decreases CTTN ability to associate with filamentous actin. Phosphorylates HDAC5, therefore lead to impair HDAC5 import. Direct RhoA target required for the regulation of the maturation of primordial junctions into apical junction formation in bronchial epithelial cells. Required for G2/M phases of the cell cycle progression and abscission during cytokinesis in a ECT2-dependent manner. Stimulates FYN kinase activity that is required for establishment of skin cell-cell adhesion during keratinocytes differentiation. Regulates epithelial bladder cells speed and direction of movement during cell migration and tumor cell invasion. Inhibits Akt pro-survival-induced kinase activity. Mediates Rho protein-induced transcriptional activation via the c-fos serum response factor (SRF). Phosphorylates HCV NS5B leading to stimulation of HCV RNA replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKN2
Applications ELISA, Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone 11H3
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PKN2
Gene Accession No. Q16513
Gene Alias 6030436C20Rik; AI507382; cardiolipin-activated protein kinase Pak2; p140 kinase; PAK2; Pak-2; PKN gamma; PKN2; PRK2; PRKCL2; PRO2042; protease-activated kinase 2; protein kinase C-like 2; protein kinase N2; protein-kinase C-related kinase 2; RP5-905H16.1; serine/threonine kinase 7; serine/threonine-protein kinase N2; serine/threonine-protein kinase N2; serine/threonine-protein kinase N2-like; si:ch211-83f14.1; Stk7; wu:fa97c07
Gene Symbols PKN2
Host Species Rabbit
Immunogen A synthesized peptide derived from Human PKN2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5586
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.