Learn More
PKN3 Monoclonal Antibody (OTI11D8)

Description
PKN3 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | PKN3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI11D8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PKN3 |
| Gene Accession No. | Q6P5Z2 |
| Gene Alias | AW209115; BC034126; Pkn3; PKNBETA; protein kinase N3; protein kinase PKNbeta; protein kinase PKN-beta; protein-kinase C-related kinase 3; Serine/threonine-protein kinase N3; UTDP4-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.