Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA130299
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA130299 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA130299 Supplier Invitrogen™ Supplier No. PA130299
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa cell lysate. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PKN3, a protein kinase C-related molecule, mediates malignant cell growth downstream of activated PI3K. PKN3 is required for invasive prostate cell growth as assessed by 3D cell culture assays and in an orthotopic mouse tumor model by inducible expression of short hairpin RNA (shRNA). PKN3 is regulated by PI3K at both the expression level and the catalytic activity level. Therefore, PKN3 might represent a preferred target for therapeutic intervention in cancers that lack tumor suppressor PTEN function or depend on chronic activation of PI3K.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKN3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene PKN3
Gene Accession No. Q6P5Z2
Gene Alias AW209115; BC034126; Pkn3; PKNBETA; protein kinase N3; protein kinase PKNbeta; protein kinase PKN-beta; protein-kinase C-related kinase 3; Serine/threonine-protein kinase N3; UTDP4-1
Gene Symbols PKN3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 463-492 amino acids from the C-terminal region of human PKN beta.
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29941
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.