Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKNOX1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219853
Description
PKNOX1 Polyclonal specifically detects PKNOX1 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
PKNOX1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
Homeobox protein PREP-1, PBX/kn10Pbx regulating protein-1, pkonx1c | |
Rabbit | |
48 kDa | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
5316 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, ChIP Assay | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation -Target validated by Mass Spectrometry, Chromatin Immunoprecipitation (ChIP) -Assay dependent | |
P55347 | |
PKNOX1 | |
Recombinant protein encompassing a sequence within the center region of human PKNOX1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction