Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKNOX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566739
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566739 Supplier Invitrogen™ Supplier No. PA566739
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLSSDGATLAV QQVMMAGQSE DESVDSTEED AGALAPAHIS GLVLENSDSL Q Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.

PKNOX1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKNOX1
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pknox1
Gene Accession No. P55347
Gene Alias D17Wsu76e; homeobox protein PKNOX1; Homeobox protein PREP-1; human homeobox-containing protein; Pbx regulating protein-1; Pbx/knotted 1 homeobox; PBX/knotted 1 homeobox 1; PBX/knotted homeobox 1; PKNOX1; pkonx1c; PREP1
Gene Symbols Pknox1
Host Species Rabbit
Immunogen Recombinant Human PKNOX1. Recombinant protein control fragment (Product #RP-107395).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5316
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.