Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKNOX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590359
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590359 Supplier Invitrogen™ Supplier No. PA590359
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDSSCSETPK TKKKTAQNRP VQRFWPDSIA SGVAQPPPSE LTMSEGAVVT ITTPVNMNVD SLQSLSSDGA TLAVQQVMMA GQSEDESVDS TEEDAGALAP AHISGLVLEN SDSLQ; Positive Samples: 293T; Cellular Location: Nucleus.

PKNOX1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKNOX1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Pknox1
Gene Accession No. O70477, P55347
Gene Alias D17Wsu76e; homeobox protein PKNOX1; Homeobox protein PREP-1; human homeobox-containing protein; Pbx regulating protein-1; Pbx/knotted 1 homeobox; PBX/knotted 1 homeobox 1; PBX/knotted homeobox 1; PKNOX1; pkonx1c; PREP1
Gene Symbols Pknox1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 322-436 of human PKNOX1 (NP_0045622).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18771, 5316
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.