Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKNOX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565032
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565032 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565032 Supplier Invitrogen™ Supplier No. PA565032
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEKVNELCKDF CNRYITCLKT KMHSDNLLRN DLGGPYSPNQ PSINLHSQDL LQNSPNSMSG VSNNPQGIVV PASALQQGNI AMTTVNSQVV SGGA Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

PREP-2 (Pbx-regulating protein-2), also known as PBX/knotted 1 homeobox 2 or PKNOX2, is a widely expressed protein belonging to the TALE (three amino acid loop extension)/MEIS family. PREP-2 is a DNA-binding protein that forms stable complexes with Pbx proteins. It is highly homologous to the related protein PREP-1, but displays a more restricted tissue distribution and a higher DNA-dissociation rate. Like PREP-1, PREP-2 forms a heterodimer with Pbx 1. The PREP-2-Pbx 1 dimer is relocated to the nucleus where it associates with HoxB1 to form a ternary complex. In contrast with PREP-1, which acts to increase transcriptional activation in this ternary complex, PREP-2 leads to a slight decrease in transcriptional activity of the ternary complex. Multiple isoforms exist for PREP-2, localizing to the nucleus or cytoplasm. Cytoplasmic isoforms are believed to colocalize with F-actin, G-actin and tubulin/microtubules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKNOX2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PKNOX2
Gene Accession No. Q96KN3
Gene Alias D230005H23Rik; homeobox protein Meis2-like; homeobox protein PKNOX2; Homeobox protein PREP-2; homeodomain containing transcription factor PREP2; Pbx/knotted 1 homeobox 2; PBX/knotted homeobox 2; Pknox2; PREP2
Gene Symbols PKNOX2
Host Species Rabbit
Immunogen Recombinant Human PKNOX2. Recombinant protein control fragment (Product #RP-105798).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63876
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.