Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKP1 Monoclonal Antibody (10B2)
GREENER_CHOICE

Catalog No. 325700
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-570-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-570-0 Supplier Invitrogen™ Supplier No. 325700
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

PKP1 Monoclonal Antibody for Western Blot, ICC/IF, IP

This gene encodes a member of the arm-repeat (armadillo) and plakophilin gene families. Plakophilin proteins contain numerous armadillo repeats, localize to cell desmosomes and nuclei, and participate in linking cadherins to intermediate filaments in the cytoskeleton. This protein may be involved in molecular recruitment and stabilization during desmosome formation. Mutations in this gene have been associated with the ectodermal dysplasia/skin fragility syndrome. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKP1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 10B2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene PKP1
Gene Accession No. Q13835
Gene Alias AI528551; armadillo; B6P; band 6; band 6 polypeptide B6P; band 6 protein; Band-6 protein; MGC138829; Pkp1; plakophilin; plakophilin 1; plakophilin 1 (ectodermal dysplasia/skin fragility syndrome); plakophilin-1
Gene Symbols PKP1
Host Species Mouse
Immunogen N-terminus of Human plakophilin-1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5317
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.