Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PKP2 Monoclonal Antibody (8H6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA546596
This item is not returnable.
View return policy
Description
PKP2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, IP
Plakophilins 1, 2, 3 and 4 (PKP1-4) influence development and participate in linking cadherins to cytoskeletal intermediate filaments. Plakophilins 1-4 contain arm-repeat (armadillo) domains and localize to nuclei and cell desmosomes (cell-cell junctions found in suprabasal layers of stratifying epithelia that undergo mechanical stress). Plakophilin 1 mediates increases in desmosomal protein content, desmosome assembly and regulation of cell migration. Plakophilin 2 is important for desmosome assembly and is an essential morphogenic factor and architectural component of the heart. Plakophilin 4 is a component of desmosomal adhesion plaques that regulates junctional plaque organization and cadherin function.
Specifications
PKP2 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q99959 | |
PKP2 | |
Recombinant fragment, corresponding to amino acids 1-350 of Human Plakophilin 2 fused to maltose binding protein. | |
50 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
8H6 | |
Unconjugated | |
PKP2 | |
1200008D14Rik; 1200012P04Rik; AA516617; ARVD9; PKP2; Pkp2l; plakophilin 2; plakophilin-2 | |
Mouse | |
Protein A | |
RUO | |
5318, 67451 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction