Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553144
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553144 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553144 Supplier Invitrogen™ Supplier No. PA553144
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGNGNLHRTS SVPEYVYNLH LVENDFVGGR SPVPKTYDML KAGTTATYEG RWGRGTAQYS SQKSVEERSL RHPLRRLEIS PDSSPERAHY THSDYQYSQR SQAGHTLHHQ ESRRAALLVP PRY Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 41%.

Plakophilins 1, 2, 3 and 4 (PKP1-4) influence development and participate in linking cadherins to cytoskeletal intermediate filaments. Plakophilins 1-4 contain arm-repeat (armadillo) domains and localize to nuclei and cell desmosomes (cell-cell junctions found in suprabasal layers of stratifying epithelia that undergo mechanical stress). Plakophilin 1 mediates increases in desmosomal protein content, desmosome assembly and regulation of cell migration. Plakophilin 2 is important for desmosome assembly and is an essential morphogenic factor and architectural component of the heart. Plakophilin 4 is a component of desmosomal adhesion plaques that regulates junctional plaque organization and cadherin function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PKP2
Gene Accession No. Q99959
Gene Alias 1200008D14Rik; 1200012P04Rik; AA516617; ARVD9; PKP2; Pkp2l; plakophilin 2; plakophilin-2
Gene Symbols PKP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PKP2. Recombinant protein control fragment (Product #RP-89628).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5318
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.