Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PKP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576614
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Plakophilins 1, 2, 3 and 4 (PKP1-4) influence development and participate in linking cadherins to cytoskeletal intermediate filaments. Plakophilins 1-4 contain arm-repeat (armadillo) domains and localize to nuclei and cell desmosomes (cell-cell junctions found in suprabasal layers of stratifying epithelia that undergo mechanical stress). Plakophilin 1 mediates increases in desmosomal protein content, desmosome assembly and regulation of cell migration. Plakophilin 2 is important for desmosome assembly and is an essential morphogenic factor and architectural component of the heart. Plakophilin 4 is a component of desmosomal adhesion plaques that regulates junctional plaque organization and cadherin function.
Specifications
PKP2 | |
Polyclonal | |
Unconjugated | |
PKP2 | |
1200008D14Rik; 1200012P04Rik; AA516617; ARVD9; PKP2; Pkp2l; plakophilin 2; plakophilin-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
287925, 5318, 67451 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99959 | |
PKP2 | |
Recombinant full length Human PKP2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction