Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKP3 Monoclonal Antibody (7F6)
GREENER_CHOICE

Catalog No. PIMA518080
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518080 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518080 Supplier Invitrogen™ Supplier No. MA518080
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This target has a predicted molecular weight of 87 kDa.

Plakophilin-3 is an Armadillo-like protein present in nuclei and desmosomes of epithelial cells. The expression pattern of this protein seems to be largely restricted to epithelial cell types. Plakophilin-3 can be detected along cell borders in a punctuate staining pattern typical for desmosomal proteins. In addition to the desmosomal immunolocalization, immunostaining was observed as bright nuclear speckles. Thus, like plakophilin-1 and-2, plakophilin-3 displays a dual intracellular localization in the desmosomal plaque and in the cell nucleus, and therefore is probably involved in signal transduction pathways between the plasma membrane and the nucleus. The human protein has a predicted molecular mass of ~87 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKP3
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene PKP3
Gene Accession No. Q9QY23, Q9Y446
Gene Alias 2310056L12Rik; Pkp3; plakophilin 3; plakophilin 3b; plakophilin-3
Gene Symbols PKP3
Host Species Mouse
Immunogen Recombinant fragment corresponding to amino acids 1-308 of Human Plakophilin 3 fused to maltose binding protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11187, 56460
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.