Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101752
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101752 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101752 Supplier Invitrogen™ Supplier No. PA5101752
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PKP4.

Plays a role as a regulator of Rho activity during cytokinesis. May play a role in junctional plaques.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKP4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PKP4
Gene Accession No. Q68FH0, Q99569
Gene Alias 5031422I09Rik; 9430019K17Rik; Armadillo-related protein; Armrp; catenin 4; p0071; PKP4; plakophilin 4; plakophilin-4
Gene Symbols PKP4
Host Species Rabbit
Immunogen A synthesized peptide derived from human PKP4(Accession Q99569), corresponding to amino acid residues T184-R234.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227937, 295625, 8502
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.