Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PKP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA535286
Description
PKP4 Polyclonal Antibody for Western Blot, IHC (P)
Plays a role as a regulator of Rho activity during cytokinesis. May play a role in junctional plaques.
Specifications
| PKP4 | |
| Polyclonal | |
| Unconjugated | |
| PKP4 | |
| 5031422I09Rik; 9430019K17Rik; Armadillo-related protein; Armrp; catenin 4; p0071; PKP4; plakophilin 4; plakophilin-4 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 227937, 8502 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q68FH0, Q99569 | |
| PKP4 | |
| KLH conjugated synthetic peptide between 1040-1068 amino acids from the C-terminal region of human PKP4. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction