Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKR, p Thr446 Antibody (SR2125), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32152150UL
Description
PKR Monoclonal antibody specifically detects PKR in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
PKR | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
SR2125 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
double stranded RNA activated protein kinase, EC 2.7.11.1, eIF-2A protein kinase 2, EIF2AK1, eukaryotic translation initiation factor 2-alpha kinase 2P1/eIF-2A protein kinase, interferon-induced, double-stranded RNA-activated protein kinase, interferon-inducible double stranded RNA dependent, interferon-inducible elF2alpha kinase, Interferon-inducible RNA-dependent protein kinase, PKRp68 kinase, PRKRMGC126524, Protein kinase RNA-activated | |
A synthesized peptide derived from human phospho-PKR [Thr446] (Uniprot #: P19525) | |
50 μg | |
Apoptosis, Phospho Specific, Signal Transduction | |
5610 | |
Store at -20 degrees C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction