Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566277
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566277 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566277 Supplier Invitrogen™ Supplier No. PA566277
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QCQINQVKFKF QSSNRVWKKD RTTIIGKFCT ALLPVNDREK MVCLPEPVNL QASVTVSCDL KIAC Highest antigen sequence identity to the following orthologs - mouse 71%, rat 68%.

PLA1A is a phospholipase that hydrolyzes fatty acids at the sn-1 position of phosphatidylserine and 1-acyl-2-lysophosphatidylserine. This secreted protein hydrolyzes phosphatidylserine (PS) in liposomes and can also hydrolyze PS in apoptotic cells and activate platelets where the resulting 2-acyl-lysophosphatidylserine acts as a lipid mediator for mast cells, T cells, and neural cells, suggesting that a major function of PLA1A may be the production of lysophospholipid mediators. PLA1A is upregulated in rat peripheral blood cells bearing long-term surviving cardiac allograft. PLA1A is also expressed in human THP-1-derived macrophages and this expression is upregulated in cells treated with lipopolysaccharide, a TLR4 ligand. This upregulation is inhibited with corticosteroids, which are often used at high dosages to suppress chronic allograft rejection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA1A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLA1A
Gene Accession No. Q53H76
Gene Alias AA986889; NMD; Phosphatidylserine-specific phospholipase A1; phosphatidylserine-specific phospholipase A1alpha; phospholipase A1 member A; PLA1A; PSPLA1; PS-PLA1
Gene Symbols PLA1A
Host Species Rabbit
Immunogen Recombinant Human PLA1A. Recombinant protein control fragment (Product #RP-106963).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51365
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.