Learn More
Invitrogen™ PLA2G10 Polyclonal Antibody

Description
Immunogen sequence: ASRILRVHRR GILELAGTVG CVGPRTPIAY MKYGCFCGLG GHGQPRDAID WCCH Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 72%.
Specifications
Specifications
| Antigen | PLA2G10 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PLA2G10 |
| Gene Accession No. | O15496 |
| Gene Alias | group 10 secretory phospholipase A2; group 10 secretory phospholipase A2-like; group X secretory phospholipase A2; GX sPLA2; GXPLA2; GXSPLA2; LOC100652777; Phosphatidylcholine 2-acylhydrolase 10; phospholipase A2 group X; phospholipase A2, group X; PLA2G10; SPLA2; sPLA2-X |
| Gene Symbols | PLA2G10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.