Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA2G12A Monoclonal Antibody (3H2C11)
GREENER_CHOICE

Catalog No. PIMA531851
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531851 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531851 Supplier Invitrogen™ Supplier No. MA531851
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31851 has been tested in indirect ELISA.

Secreted phospholipase A2 (sPLA2) enzymes liberate arachidonic acid from phospholipids for production of eicosanoids and exert a variety of physiologic and pathologic effects. Group XII sPLA2s, such as PLA2G12A, have relatively low specific activity and are structurally and functionally distinct from other sPLA2s.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G12A
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3H2C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PLA2G12A
Gene Accession No. Q9BZM1
Gene Alias 2310004B05Rik; FKSG38; group XII secreted phospholipase A2; group XII-1 phospholipase A2; group XIIA secreted phospholipase A2; group XIIA secretory phospholipase A2; GXII; GXII sPLA2; GXII-1; GXII-1-PLA2; mGXII-1; phosphatidylcholine 2-acylhydrolase 12A; phospholipase A2 group XIIA; phospholipase A2, group XII; phospholipase A2, group XIIA; Pla2g12; PLA2G12A; Rossy; sPLA2-XII; UNQ2519/PRO6012
Gene Symbols PLA2G12A
Host Species Mouse
Immunogen Purified recombinant fragment of human PLA2G12A (AA: 21-189) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81579
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.