Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLA2G12A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530867
Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (97%), Xenopus laevis (83%), Rhesus Monkey (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Secreted phospholipase A2 (sPLA2) enzymes liberate arachidonic acid from phospholipids for production of eicosanoids and exert a variety of physiologic and pathologic effects. Group XII sPLA2s, such as PLA2G12A, have relatively low specific activity and are structurally and functionally distinct from other sPLA2s.
Specifications
| PLA2G12A | |
| Polyclonal | |
| Unconjugated | |
| PLA2G12A | |
| 2310004B05Rik; FKSG38; group XII secreted phospholipase A2; group XII-1 phospholipase A2; group XIIA secreted phospholipase A2; group XIIA secretory phospholipase A2; GXII; GXII sPLA2; GXII-1; GXII-1-PLA2; mGXII-1; phosphatidylcholine 2-acylhydrolase 12A; phospholipase A2 group XIIA; phospholipase A2, group XII; phospholipase A2, group XIIA; Pla2g12; PLA2G12A; Rossy; sPLA2-XII; UNQ2519/PRO6012 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81579 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.84 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9BZM1 | |
| PLA2G12A | |
| Recombinant protein encompassing a sequence within the center region of human Phospholipase A2 XIIA. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction