Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLA2G16/HRASLS3 Antibody (OTI1A5) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273454
Description
PLA2G16/HRASLS3 Monoclonal specifically detects PLA2G16/HRASLS3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
PLA2G16/HRASLS3 | |
Monoclonal | |
LYOPH | |
Western Blot 1:4000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100 | |
Adipose-specific phospholipase A2, AdPLAHRASLS3, EC 3.1.1.4, group XVI phospholipase A2, HRAS-like suppressor 3Ca-independent phospholipase A1/2, H-REV107-1, HREV107-3adipose-specific PLA2, HREV107H-rev 107 protein homolog, MGC118754, phospholipase A2, group XVI, Renal carcinoma antigen NY-REN-65 | |
Mouse | |
17.8 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1A5 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
PLA2G16 | |
Full length human recombinant protein of human PLA2G16(NP_009000) produced in HEK293T cell. | |
Affinity Purified | |
Stem Cell Markers | |
11145 | |
Human | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction