Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLA2G16 Monoclonal Antibody (OTI1B12), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

$516.29

Specifications

Antigen PLA2G16
Clone OTI1B12
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-167-6850
SDP
Encompass_Preferred
View Documents
OriGene
TA506914
100 μL
Each of 1 for $516.29
Only null left
Add to Cart
 
Description

Description

PLA2G16 Monoclonal antibody specifically detects PLA2G16 in Human samples. It is validated for Western Blot
Specifications

Specifications

PLA2G16
1 mg/mL
Monoclonal
Liquid
RUO
P53816
IgG1
-20° C, Avoid Freeze/Thaw Cycles
OTI1B12
Western Blot
Unconjugated
Mouse
Human
11145
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.