Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLA2G16 Monoclonal Antibody (OTI1H1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA506909

Description
HRASLS3 specifically catalyzes the release of fatty acids from phospholipids in adipose tissue. It also has a weak lysophospholipase activity. Tumor suppressor that may be involved in interferon-dependent cell death.Specifications
PLA2G16 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P53816 | |
PLA2G16 | |
Full length human recombit protein of human PLA2G16 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI1H1 | |
Unconjugated | |
PLA2G16 | |
AdPLA, H-REV107, H-REV107-1, HRASLS3, HREV107, HREV107-1, HREV107-3, HRSL3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
11145 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction