Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLA2G1B Antibody (001), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290537UV
Description
PLA2G1B Monoclonal specifically detects PLA2G1B in Mouse samples. It is validated for ELISA.Specifications
PLA2G1B | |
Monoclonal | |
DyLight 350 | |
Group IB phospholipase A2, Phosphatidylcholine 2-acylhydrolase 1B, phospholipase A2, phospholipase A2, group IB (pancreas), PLA2AMGC119834, PLA2EC 3.1.1.4, PPLA2MGC119835 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse PLA2G1B (NP_035237.1; Met 1-Cys146). | |
0.1 mL | |
Lipid and Metabolism | |
5319 | |
Mouse | |
Purified |
ELISA | |
1 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human PLA2G1B | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction