Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLA2G1B Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298077AF350
Description
PLA2G1B Polyclonal specifically detects PLA2G1B in Mouse samples. It is validated for ELISA.Specifications
PLA2G1B | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
Alexa Fluor 350 | |
Group IB phospholipase A2, Phosphatidylcholine 2-acylhydrolase 1B, phospholipase A2, phospholipase A2, group IB (pancreas), PLA2AMGC119834, PLA2EC 3.1.1.4, PPLA2MGC119835 | |
Produced in rabbits immunized with purified, recombinant PLA2G1B (NP_035237.1; Met 1-Cys146). | |
0.1 mL | |
Lipid and Metabolism | |
5319 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction