Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLA2G1B Mouse anti-Human, Porcine, Clone: 1F2C10, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | PLA2G1B |
---|---|
Clone | 1F2C10 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6825
![]() ![]() |
Proteintech Group Inc
663971IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Phospholipase A2 catalyzes the release of fatty acids from glycero-3-phosphocholines. The best known varieties are the digestive enzymes secreted as zymogens by the pancreas of mammals. Sequences of pancreatic PLA2 enzymes from a variety of mammals have been reported. One striking feature of these enzymes is their close homology to venom phospholipases of snakes. Other forms of PLA2 have been isolated from brain, liver, lung, spleen, intestine, macrophages, leukocytes, erythrocytes, inflammatory exudates, chondrocytes, and platelets.Specifications
PLA2G1B | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Group IB phospholipase A2, Phospholipase A2, PLA2, PLA2A, PLA2G1B, PPLA2 | |
PLA2G1B | |
IgG2a | |
Protein A | |
Antibody |
1F2C10 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig | |
P00592, P04054 | |
445525, 5319 | |
PLA2G1B Fusion Protein Ag8812 | |
Primary | |
-20°C | |
PLA2G1B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title