Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLA2G2A Antibody (OTI1C2), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273455AF700
Description
PLA2G2A Monoclonal specifically detects PLA2G2A in Human samples. It is validated for Western Blot.Specifications
PLA2G2A | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PLA2G2A | |
Recombinant protein fragment corresponding to amino acids 21-144 of human PLA2G2A (NP_000291) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI1C2 | |
Western Blot | |
EC 3.1.1.4, GIIC sPLA2, Group IIA phospholipase A2, MOM1, Non-pancreatic secretory phospholipase A2, NPS-PLA2, Phosphatidylcholine 2-acylhydrolase 2A, phospholipase A2, group IIA (platelets, synovial fluid), phospholipase A2, membrane associated, PLA2BPLA2, PLA2LPLA2S, PLAS1, RASF-A, sPLA2 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
5320.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction