Learn More
Invitrogen™ PLA2G2A Polyclonal Antibody

Description
Immunogen sequence: NLVNFHRMIK LTTGKEAALS YGFYGCHCGV GGRGSPKDAT DRCCVTHDCC YKRLEKRGCG TKFLSYKFSN SGSRITCAKQ DSCRSQLCEC DKAAATCFAR NKTTYNKKYQ YYSNKHCRGS TPRC.
Specifications
Specifications
| Antigen | PLA2G2A |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.28 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PLA2G2A |
| Gene Accession No. | P14555 |
| Gene Alias | EC 3.1.1.4; EF; Enhancing factor; eted enzyme type IIA phospholipase A2; GIIC sPLA2; Group IIA phospholipase A2; modifier of Min1; Mom1; non-pancreatic secreted type II phospholipase A2; non-pancreatic secretory phospholipase A2; NPS-PLA2; phosphatidylcholine 2-acylhydrolase 2A; phospholipase A2 group IIA; phospholipase A2, group IIA (platelets, synovial fluid); phospholipase A2, membrane associated; Pla2; PLA2B; PLA2BPLA2; PLA2G2A; PLA2L; PLA2LPLA2S; PLA2S; PLAS1; platelet phospholipase A2; RASF-A; secretory group II phospholipase A2; sPLA2; sPla2-IIA |
| Gene Symbols | PLA2G2A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.