Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLA2G2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576118
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The protein encoded by this gene is a member of the phospholipase A2 family (PLA2). PLA2s constitute a diverse family of enzymes with respect to sequence, function, localization, and divalent cation requirements. This gene product belongs to group II, which contains secreted form of PLA2, an extracellular enzyme that has a low molecular mass and requires calcium ions for catalysis. It catalyzes the hydrolysis of the sn-2 fatty acid acyl ester bond of phosphoglycerides, releasing free fatty acids and lysophospholipids, and thought to participate in the regulation of the phospholipid metabolism in biomembranes. Several alternatively spliced transcript variants with different 5' UTRs have been found for this gene.
Specifications
PLA2G2A | |
Polyclonal | |
Unconjugated | |
PLA2G2A | |
EC 3.1.1.4; EF; Enhancing factor; eted enzyme type IIA phospholipase A2; GIIC sPLA2; Group IIA phospholipase A2; modifier of Min1; Mom1; non-pancreatic secreted type II phospholipase A2; non-pancreatic secretory phospholipase A2; NPS-PLA2; phosphatidylcholine 2-acylhydrolase 2A; phospholipase A2 group IIA; phospholipase A2, group IIA (platelets, synovial fluid); phospholipase A2, membrane associated; Pla2; PLA2B; PLA2BPLA2; PLA2G2A; PLA2L; PLA2LPLA2S; PLA2S; PLAS1; platelet phospholipase A2; RASF-A; secretory group II phospholipase A2; sPLA2; sPla2-IIA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18780, 5320 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P14555, P31482 | |
PLA2G2A | |
Recombinant protein of human PLA2G2A. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction