Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLA2G2D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577155 Supplier Thermo Scientific Supplier No. PA577155
Add to Cart
Add to Cart

PLA2G2D Polyclonal Antibody

PLA2G2D Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G2D
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Pla2g2d
Gene Accession No. Q9UNK4
Gene Alias GIID sPLA2; Group IID secretory phospholipase A2; mGIID; phosphatidylcholine 2-acylhydrolase 2D; phospholipase A2 group IID; phospholipase A2, group IID; Pla2a2; PLA2G2D; PLA2IID; secretory phospholipase A2s; secretory-type PLA, stroma-associated homolog; sPLA2-IID; sPLA2S; SPLASH
Gene Symbols Pla2g2d
Host Species Rabbit
Immunogen Recombinant full length Human PLA2G2D.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26279, 298579
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.