Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA2G2D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596597
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596597 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596597 Supplier Invitrogen™ Supplier No. PA596597
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GILNLNKMVK QVTGKMPILS YWPYGCHCGL GGRGQPKDAT DWCCQTHDCC YDHLKTQGCS IYKDYYRYNF SQGNIHCSDK GSWCEQQLCA CDKEVAFCLK RNLDTYQKRL RFYWRPHCRG QTPGC; Positive Samples: Rat lung, Rat spinal cord.

PA2 catalyzes the calcium-dependent hydrolysis of the 2- acyl groups in 3-sn-phosphoglycerides. L-alpha-1-palmitoyl-2- linoleoyl phosphatidylethanolamine is more efficiently hydrolyzed than the other phospholipids examined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G2D
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Pla2g2d
Gene Accession No. Q9UNK4
Gene Alias GIID sPLA2; Group IID secretory phospholipase A2; mGIID; phosphatidylcholine 2-acylhydrolase 2D; phospholipase A2 group IID; phospholipase A2, group IID; Pla2a2; PLA2G2D; PLA2IID; secretory phospholipase A2s; secretory-type PLA, stroma-associated homolog; sPLA2-IID; sPLA2S; SPLASH
Gene Symbols Pla2g2d
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26279, 298579
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.