Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA2G2E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564150
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564150 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564150 Supplier Invitrogen™ Supplier No. PA564150
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GNLVQFGVMI EKMTGKSALQ YNDYGCYCGI G Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 56%.

Secretory calcium-dependent phospholipase A2 that primarily targets extracellular phospholipids (PubMed:10681567, PubMed:11922621, PubMed:28883454). Hydrolyzes the ester bond of the fatty acyl group attached at sn-2 position of phospholipids (phospholipase A2 activity), releasing various unsaturated fatty acids including oleoate, linoleoate, arachidonate, docosahexaenoate and lysophosphatidylethanolamines in preference to lysophosphatidylcholines (PubMed:10681567, PubMed:28883454). In response to high-fat diet, hydrolyzes minor lipoprotein phospholipids including phosphatidylserines, phosphatidylinositols and phosphatidylglycerols, altering lipoprotein composition and fat storage in adipose tissue and liver. May act in an autocrine and paracrine manner (PubMed:11922621). Contributes to lipid remodeling of cellular membranes and generation of lipid mediators involved in pathogen clearance. Cleaves sn-2 fatty acyl chains of phosphatidylglycerols and phosphatidylethanolamines, which are major components of membrane phospholipids in bacteria (PubMed:11922621). Acts as a hair follicle phospholipase A2. Selectively releases lysophosphatidylethanolamines (LPE) and various unsaturated fatty acids in skin to regulate hair follicle homeostasis. May regulate the inflammatory response by releasing arachidonate, a precursor of prostaglandins and leukotrienes (PubMed:11922621). Upon allergen exposure, may participate in allergic inflammatory response by enhancing leukotriene C4 synthesis and degranulation in mast cells. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G2E
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLA2G2E
Gene Accession No. Q9NZK7
Gene Alias GIIE sPLA2; Group IIE secretory phospholipase A2; mGIIEsPLA2s; phosphatidylcholine 2-acylhydrolase 2E; phospholipase A2 group IIE; phospholipase A2, group IIE; Pla2g2e; sPLA2-IIE
Gene Symbols PLA2G2E
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLA2G2E. Recombinant protein control fragment (Product #RP-103543).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30814
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.