Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLA2G3 Monoclonal Antibody (OTI4F2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA811652

Description
PLA2G3 belongs to the family of secreted phospholipase A2 (sPLA2; EC 3.1.1.4) proteins. These Ca(2+)-dependent lipolytic enzymes have a conserved Ca(2+)-binding loop and a his-asp dyad in the catalytic site.Specifications
PLA2G3 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9NZ20 | |
PLA2G3 | |
Human recombit protein fragment corresponding to amino acids 409-509 of human PLA2G3 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI4F2 | |
Unconjugated | |
PLA2G3 | |
GIII-SPLA2, SPLA2III, sPLA2-III | |
Mouse | |
Affinity Chromatography | |
RUO | |
50487 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction